Gene Transcripts

Gene Symbol
Cdk17
Entrez Gene ID
237459
Gene Name
cyclin-dependent kinase 17
Gene Aliases
6430598J10Rik, Pctk2
Chromosome Location
Chr. 10: 93160876 - 93250688 on Build GRCm38
UniGene ID
Mm.217404
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146239.2 NP_666351.2 8 1106
GenBank mRNA BC031778.1 - 653
BC049904.1 - 653 4501115
BC064815.1 - 8 1091 6516981

View Full Product Details