Gene Transcripts

Gene Symbol
Csmd3
Entrez Gene ID
239420
Gene Name
CUB and Sushi multiple domains 3
Gene Aliases
4930500N14Rik, mKIAA1894
Chromosome Location
Chr. 15: 47580633 - 48792231 on Build GRCm38
UniGene ID
Mm.309363
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081391.2 NP_001074860.2 61 9849
GenBank mRNA AK122567.1 - 44 7032

View Full Product Details