Gene Transcripts

Gene Symbol
Bloc1s2
Entrez Gene ID
73689
Gene Name
biogenesis of lysosomal organelles complex-1, subunit 2
Gene Aliases
2410089B13Rik, BLOS2, Bloc1s2a
Chromosome Location
Chr. 19: 44139246 - 44146470 on Build GRCm38
UniGene ID
Mm.124685
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028607.1 NP_082883.1 3 287
GenBank mRNA AK010745.1 - 3 284
AK145313.1 - 3 288
BC048500.1 - 2 167
BC065806.1 - 3 280 1547643

View Full Product Details