Gene Transcripts

Gene Symbol
Tekt2
Entrez Gene ID
24084
Gene Name
tektin 2
Gene Aliases
AU014974, tektin-t
Chromosome Location
Chr. 4: 126322121 - 126325199 on Build GRCm38
UniGene ID
Mm.23895
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011902.2 NP_036032.2 3 317
GenBank mRNA AB027138.1 - 3 317
AK006205.1 - 2 64
BC008140.1 - 3 333 3582971
BQ712964.1 - 3 331

View Full Product Details