Gene Transcripts

Gene Symbol
Itgb1bp2
Entrez Gene ID
26549
Gene Name
integrin beta 1 binding protein 2
Gene Aliases
Chordc3
Chromosome Location
Chr. X: 101448690 - 101466122 on Build GRCm38
UniGene ID
Mm.46232
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013712.2 NP_038740.1 4 361
GenBank mRNA AF140691.1 - 4 305
AK003906.1 - 4 362
BC030035.1 - 4 336 1346742

View Full Product Details