Gene Transcripts

Gene Symbol
Slc4a11
Entrez Gene ID
269356
Gene Name
solute carrier family 4, sodium bicarbonate transporter-like, member 11
Gene Aliases
AI503023, BTR1, NaBC1
Chromosome Location
Chr. 2: 130684107 - 130697519 on Build GRCm38
UniGene ID
Mm.441391
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081162.1 NP_001074631.1 4 411
GenBank mRNA AK052665.1 - 4 362
AK158115.1 - 4 365
BC111884.1 - 3 206 40060299

View Full Product Details