Gene Transcripts

Gene Symbol
Klhl23
Entrez Gene ID
277396
Gene Name
kelch-like 23
Gene Aliases
C130068N17Rik
Chromosome Location
Chr. 2: 69821920 - 69836651 on Build GRCm38
UniGene ID
Mm.138073
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177784.4 NP_808452.2 2 876
GenBank mRNA AK048518.1 - 2 775
BC072626.1 - 2 876 6827841

View Full Product Details