Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198899.2 | NP_942602.2 | 19 | 2269 | |
GenBank mRNA | AK050532.1 | - | 19 | 2256 | |
AK087883.1 | - | 19 | 2068 | ||
AK144381.1 | - | 19 | 2122 | ||
BC062936.1 | - | 19 | 2216 | 6838545 | |
BC068283.1 | - | 19 | 2251 | 30022247 |