Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177304.3 | NP_796278.1 | 1 | 259 | |
GenBank mRNA | AK038460.1 | - | 1 | 231 | |
AK046797.1 | - | 1 | 260 | ||
AK046881.1 | - | 1 | 264 | ||
AK079352.1 | - | 1 | 241 | ||
AK079642.1 | - | 1 | 258 | ||
BC096376.1 | - | 1 | 230 | 4237600 |