Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289809.1 | NP_001276738.1 | 10 | 1457 | |
NM_001289811.1 | NP_001276740.1 | 11 | 1577 | ||
NM_183307.4 | NP_899130.1 | 11 | 1719 | ||
GenBank mRNA | AK029513.1 | - | 11 | 1721 | |
BC118944.1 | - | 10 | 1379 | ||
BC118945.1 | - | 11 | 1499 | 40092360 |