Gene Transcripts

Gene Symbol
Trim71
Entrez Gene ID
636931
Gene Name
tripartite motif-containing 71
Gene Aliases
2610206G21Rik, AL022943, Gm1127, Lin41, lin-41, mLin41, mlin-41
Chromosome Location
Chr. 9: 114511268 - 114564369 on Build GRCm38
UniGene ID
Mm.17857
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042503.2 NP_001035968.1 4 2630
GenBank mRNA DQ005956.1 - 4 2629

View Full Product Details