Gene Transcripts

Gene Symbol
Phc2
Entrez Gene ID
54383
Gene Name
polyhomeotic-like 2 (Drosophila)
Gene Aliases
A3galt2, AA415044, D130050K19Rik, D4Ertd810e, Edr2, Mph2, p36
Chromosome Location
Chr. 4: 128654673 - 128752881 on Build GRCm38
UniGene ID
Mm.473738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195083.1 NP_001182012.1 4 666
NM_001195130.1 NP_001182059.1 12 2066
NM_018774.4 NP_061244.1 12 2059
GenBank mRNA AB062362.1 - 4 652
AF060076.1 - 5 645
BC051657.1 - 4 559
BC057571.1 - 12 2002 6410302
BC071246.1 - 4 647
U81491.1 - 11 1956

View Full Product Details