Gene Transcripts

Gene Symbol
Ylpm1
Entrez Gene ID
56531
Gene Name
YLP motif containing 1
Gene Aliases
A930013E17Rik, ZAP, Zap3
Chromosome Location
Chr. 12: 84996285 - 85074533 on Build GRCm38
UniGene ID
Mm.153183
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178363.3 NP_848140.3 9, 10 5375
GenBank mRNA AB033168.1 - 9, 10 3048
AK162250.1 - 1, 2 84

View Full Product Details