Gene Transcripts
- Gene Symbol
- Psmd8
- Entrez Gene ID
- 57296
- Gene Name
- proteasome (prosome, macropain) 26S subunit, non-ATPase, 8
- Gene Aliases
- 6720456J22Rik, AA407360, AL033291, AL033322, AL033323, C76433
- Chromosome Location
- Chr. 7: 29174187 - 29180673 on Build GRCm38
- UniGene ID
- Mm.273152
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK013113.1 |
- |
|
183 |
|
Chr. - - [-] |
View Full Product Details