Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164466.1 | NP_001157938.1 | 2, 3 | 522 | |
NM_022722.3 | NP_073559.3 | 2, 3 | 522 | ||
GenBank mRNA | AF227731.1 | - | 2, 3 | 440 | |
AF249296.1 | - | 2, 3 | 509 | ||
AK004822.1 | - | 2, 3 | 522 | ||
AK004899.2 | - | 2, 3 | 519 | ||
BC037086.1 | - | 2, 3 | 491 | 5123860 |