Gene Transcripts

Gene Symbol
Cldn12
Entrez Gene ID
64945
Gene Name
claudin 12
Gene Aliases
Chromosome Location
Chr. 5: 5505015 - 5514976 on Build GRCm38
UniGene ID
Mm.40132
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193659.1 NP_001180588.1 4 702
NM_001193660.1 NP_001180589.1 5 650
NM_001193661.1 NP_001180590.1 4 537
NM_022890.2 NP_075028.1 4 692
GenBank mRNA AF247664.1 - 1 284
AK039672.1 - 4 584
AK049331.1 - 4 559
AK128999.1 - 4 571
AK132155.1 - 4 561
AK166110.1 - 4 513
BC024057.1 - 4 601 5100104

View Full Product Details