Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025360.2 | NP_079636.2 | 3 | 645 | |
GenBank mRNA | AK002698.1 | - | 3 | 593 | |
AK004545.1 | - | 3 | 593 | ||
AK007544.1 | - | 3 | 587 | ||
AK195044.1 | - | 1 | 26 | ||
BC023338.1 | - | 3 | 539 | 5253777 |