Gene Transcripts

Gene Symbol
Lrrk2
Entrez Gene ID
66725
Gene Name
leucine-rich repeat kinase 2
Gene Aliases
4921513O20Rik, 9330188B09Rik, AW561911, D630001M17Rik, Gm927, cI-46
Chromosome Location
Chr. 15: 91672821 - 91816124 on Build GRCm38
UniGene ID
Mm.37558
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025730.3 NP_080006.3 25 3533
GenBank mRNA AK034413.1 - 2 157
AY792512.1 - 25 3476

View Full Product Details