Gene Transcripts

Gene Symbol
Dgat2
Entrez Gene ID
67800
Gene Name
diacylglycerol O-acyltransferase 2
Gene Aliases
0610010B06Rik, ARAT, DGAT-2
Chromosome Location
Chr. 7: 99153663 - 99182713 on Build GRCm38
UniGene ID
Mm.180189
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026384.3 NP_080660.1 6 887
GenBank mRNA AF384160.1 - 6 745
AK002443.1 - 6 892
AK016660.1 - 6 836
BC043447.1 - 6 887 5125951

View Full Product Details