Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_197987.2 | NP_932104.1 | 15 | 1915 | |
GenBank mRNA | AB093271.1 | - | 15 | 1886 | |
AK029570.1 | - | 1 | 190 | ||
AK086798.1 | - | 2 | 290 | ||
AK140822.1 | - | 15 | 1916 | ||
BC022117.1 | - | 10 | 1081 | ||
BC058678.1 | - | 15 | 1913 | 6834513 | |
BC059070.1 | - | 15 | 1881 | 6825999 | |
BC078416.1 | - | 15 | 1900 |