Gene Transcripts

Gene Symbol
Ttc39c
Entrez Gene ID
72747
Gene Name
tetratricopeptide repeat domain 39C
Gene Aliases
1700008N02Rik, 2810439F02Rik
Chromosome Location
Chr. 18: 12643533 - 12737052 on Build GRCm38
UniGene ID
Mm.52526
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028341.4 NP_082617.2 12 1892
GenBank mRNA AK013267.1 - 12 1627
AK077971.1 - 952

View Full Product Details