Gene Transcripts

Gene Symbol
Sass6
Entrez Gene ID
72776
Gene Name
spindle assembly 6 homolog (C. elegans)
Gene Aliases
2810453L12Rik
Chromosome Location
Chr. 3: 116594958 - 116630986 on Build GRCm38
UniGene ID
Mm.440836
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289568.1 NP_001276497.1 5 468
NM_001289571.1 NP_001276500.1 3 324
NM_028349.3 NP_082625.2 4 385
GenBank mRNA AK013338.1 - 5 360
AK145076.1 - 3 273
AK165224.1 - 5 414
BB617231.1 - 4 387
BC050110.1 - 4 335 30024366

View Full Product Details