Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023699.3 | NP_076188.3 | 9, 10 | 2880 | |
GenBank mRNA | AK159078.1 | - | 9, 10 | 2726 | |
AW493556.1 | - | 1, 2 | 401 | ||
BC028928.1 | - | 9, 10 | 2667 | 3670623 | |
EU887656.1 | - | 9, 10 | 2638 | ||
EU887658.1 | - | 4, 5 | 1093 | ||
EU887660.1 | - | 3, 4 | 2149 |