Gene Transcripts

Gene Symbol
Prss46
Entrez Gene ID
74306
Gene Name
protease, serine 46
Gene Aliases
1700112C13Rik, 79H19B, Tessp6
Chromosome Location
Chr. 9: 110844506 - 110856522 on Build GRCm38
UniGene ID
Mm.87419
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183103.2 NP_898926.2 3 509
GenBank mRNA AK007173.1 - 3 508
BC087878.1 - 2 466 30284660

View Full Product Details