Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028868.3 | NP_083144.1 | 14 | 1984 | |
GenBank mRNA | AK010337.1 | - | 14 | 1985 | |
AK083655.1 | - | 14 | 1989 | ||
AK146688.1 | - | 14 | 2068 | ||
AY099096.1 | - | 14 | 1956 | ||
BC012261.1 | - | 10 | 1527 | ||
BC030938.1 | - | 14 | 1816 | 3980339 |