Gene Transcripts

Gene Symbol
Jakmip1
Entrez Gene ID
76071
Gene Name
janus kinase and microtubule interacting protein 1
Gene Aliases
5830437M04Rik, C330021K24Rik, Gababrbp, Marlin-1
Chromosome Location
Chr. 5: 37028282 - 37150614 on Build GRCm38
UniGene ID
Mm.85280
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178394.4 NP_848481.2 7 1469
GenBank mRNA AK077298.1 - 7 1411
BC138646.1 - 6 1321 8861666
BC138649.1 - 6 1321 8861669

View Full Product Details