Gene Transcripts
- Gene Symbol
- Ttyh3
- Entrez Gene ID
- 78339
- Gene Name
- tweety homolog 3 (Drosophila)
- Gene Aliases
- 2900029G13Rik, AI414930, TTY3, mTTY3
- Chromosome Location
- Chr. 5: 140620578 - 140649031 on Build GRCm38
- UniGene ID
- Mm.28947
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK052340.1 |
- |
2 |
372 |
|
View Full Product Details