Gene Transcripts
- Gene Symbol
- Mrpl43
- Entrez Gene ID
- 94067
- Gene Name
- mitochondrial ribosomal protein L43
- Gene Aliases
- 4930442D21Rik, bMRP36a
- Chromosome Location
- Chr. 19: 45005014 - 45006442 on Build GRCm38
- UniGene ID
- Mm.44174
- Species
- Mus musculus
- Gene Symbol
- Peo1
- Entrez Gene ID
- 226153
- Gene Name
- progressive external ophthalmoplegia 1 (human)
- Gene Aliases
- D19Ertd626e, PEO, Twinl
- Chromosome Location
- Chr. 19: 45006079 - 45012762 on Build GRCm38
- UniGene ID
- Mm.105585
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054159.1 |
- |
1 |
43 |
|
View Full Product Details