Gene Transcripts

Gene Symbol
Sfxn3
Entrez Gene ID
94280
Gene Name
sideroflexin 3
Gene Aliases
Chromosome Location
Chr. 19: 45047508 - 45056383 on Build GRCm38
UniGene ID
Mm.36169
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178012.1 NP_001171483.1 6 908
NM_053197.4 NP_444427.1 7 1007
GenBank mRNA AF325262.1 - 7 771
AK034514.1 - 7 724
AK089985.1 - 6 669
BC012208.1 - 7 909 3985155

View Full Product Details