Gene Transcripts

Gene Symbol
Cyp4a14
Entrez Gene ID
13119
Gene Name
cytochrome P450, family 4, subfamily a, polypeptide 14
Gene Aliases
AI314743
Chromosome Location
Chr. 4: 115486200 - 115496159 on Build GRCm38
UniGene ID
Mm.250901
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007822.2 NP_031848.1 9 1165
GenBank mRNA AK098088.1 - 9 1162
AK149392.1 - 9 1163
AK165452.1 - 9 1163
BC089609.1 - 9 1165

View Full Product Details