Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010112.4 | NP_034242.2 | 4 | 2408 | |
GenBank mRNA | AK030321.1 | - | 4 | 1535 | |
AK163191.1 | - | 4 | 1449 | ||
BC005438.1 | - | 4 | 1477 | 3487600 | |
BC031543.1 | - | 4 | 1203 | 3669513 | |
U28728.1 | - | 4 | 1432 | ||
U57686.1 | - | 4 | 1132 |