Gene Transcripts

Gene Symbol
Hoxa13
Entrez Gene ID
15398
Gene Name
homeobox A13
Gene Aliases
Hd, Hox-1.10
Chromosome Location
Chr. 6: 52258853 - 52260880 on Build GRCm38
UniGene ID
Mm.377086
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008264.1 NP_032290.1 1 151
GenBank mRNA BC100731.1 - 1, 2 73
BC100732.1 - 1 73 40045368
BC100733.1 - 1 73 40045370

View Full Product Details