Gene Transcripts
- Gene Symbol
- Vmn1r16
- Entrez Gene ID
- 171202
- Gene Name
- vomeronasal 1 receptor 16
- Gene Aliases
- V1rc29
- Chromosome Location
- Chr. 6: 57322724 - 57323635 on Build GRCm38
- UniGene ID
- Mm.431971
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134184.2 |
NP_598945.2
|
1 |
888 |
|
View Full Product Details