Gene Transcripts

Gene Symbol
Pdgfa
Entrez Gene ID
18590
Gene Name
platelet derived growth factor, alpha
Gene Aliases
Chromosome Location
Chr. 5: 138976381 - 138999916 on Build GRCm38
UniGene ID
Mm.2675
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008808.3 NP_032834.1 2 327
GenBank mRNA AY324648.1 - 2 316
BC003817.1 - 2 327 3495629
M29464.1 - 2 207

View Full Product Details