Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286865.1 | NP_001273794.1 | 4 | 522 | |
NM_011275.3 | NP_035405.2 | 4 | 522 | ||
GenBank mRNA | AF048993.1 | - | 4 | 483 | |
AK088032.1 | - | 2 | 1037 | ||
BC019411.1 | - | 4 | 451 | 5133942 | |
BG084607.2 | - | 1 | 31 | ||
BM943446.1 | - | 1 | 42 | ||
CA554498.1 | - | 4 | 473 |