Gene Transcripts

Gene Symbol
Cd40lg
Entrez Gene ID
21947
Gene Name
CD40 ligand
Gene Aliases
CD154, CD40-L, Cd40l, HIGM1, IGM, IMD3, Ly-62, Ly62, T-BAM, TRAP, Tnfsf5, gp39
Chromosome Location
Chr. X: 57212143 - 57224042 on Build GRCm38
UniGene ID
Mm.4861
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011616.2 NP_035746.2 5 762
GenBank mRNA BC119225.1 - 5 809
EU274582.1 - 5 750
X65453.2 - 5 762

View Full Product Details