Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144559.2 | NP_653142.2 | 3, 4 | 381 | |
GenBank mRNA | AF499613.1 | - | 3, 4 | 349 | |
AK076532.1 | - | 3, 4 | 387 | ||
AK087863.1 | - | 3, 4 | 381 | ||
AK170920.1 | - | 3, 4 | 353 | ||
BC027310.1 | - | 3, 4 | 368 | 3674499 | |
EU050648.1 | - | 3, 4 | 327 |