Gene Transcripts

Gene Symbol
Spaca5
Entrez Gene ID
278203
Gene Name
sperm acrosome associated 5
Gene Aliases
Gm718
Chromosome Location
Chr. X: 21068488 - 21077959 on Build GRCm38
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085393.2 NP_001078862.1 4 419

View Full Product Details