Gene Transcripts

Gene Symbol
Tcrb-J
Entrez Gene ID
21580
Gene Name
T cell receptor beta, joining region
Gene Aliases
5830405F06Rik
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.333026
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK017898.1 - 4 432
M11456.1 - 2 253
M13676.1 - 2 226
M16679.1 - 2 223
M20877.1 - 1 175
M20878.1 - 1 175
U46842.1 - 2 226
X00438.1 - 2 248
X02778.1 - 2 202
X02779.1 - 2 226

View Full Product Details