Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175279.3 | NP_780488.2 | 1 | 249 | |
GenBank mRNA | AK031660.1 | - | 1 | 250 | |
AK046433.1 | - | 1 | 245 | ||
BB621318.1 | - | 1 | 250 | ||
BC125291.1 | - | 1 | 65 | 40129806 | |
BC125293.1 | - | 1 | 65 | 40129808 |