Gene Transcripts

Gene Symbol
Vps37c
Entrez Gene ID
107305
Gene Name
vacuolar protein sorting 37C (yeast)
Gene Aliases
5730409F24Rik, AU042646
Chromosome Location
Chr. 19: 10688729 - 10714628 on Build GRCm38
UniGene ID
Mm.19091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181403.2 NP_852068.1 2 237
GenBank mRNA AK150606.1 - 2 289
AK158833.1 - 2 148
AK159309.1 - 2 310
BC025865.1 - 2 237 5029143

View Full Product Details