Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008104.2 | NP_032130.2 | 5 | 785 | |
GenBank mRNA | AF081556.1 | - | 5 | 751 | |
AK132499.1 | - | 3 | 569 | ||
AK135689.1 | - | 7 | 784 | ||
BC110631.1 | - | 5 | 815 | 40054293 | |
BC110632.1 | - | 5 | 815 | 40054295 | |
D88611.1 | - | 5 | 992 |