Gene Transcripts

Gene Symbol
Sobp
Entrez Gene ID
109205
Gene Name
sine oculis-binding protein homolog (Drosophila)
Gene Aliases
2900009C16Rik, 5330439J01Rik, JXC1, jc
Chromosome Location
Chr. 10: 43002500 - 43174588 on Build GRCm38
UniGene ID
Mm.153750
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175407.3 NP_780616.4 3 825
GenBank mRNA BC057055.1 - 3 504
BC059851.1 - 3 825 5704623
DQ157775.1 - 3 384

View Full Product Details