Gene Transcripts
- Gene Symbol
- Sccpdh
- Entrez Gene ID
- 109232
- Gene Name
- saccharopine dehydrogenase (putative)
- Gene Aliases
- AW214504, C330023F11Rik
- Chromosome Location
- Chr. 1: 179668231 - 179687184 on Build GRCm38
- UniGene ID
- Mm.136458
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178653.3 |
NP_848768.1
|
1, 2 |
315 |
|
GenBank mRNA |
AK042919.1 |
- |
1, 2 |
203 |
|
AK046033.1 |
- |
1, 2 |
316 |
|
AK046468.1 |
- |
1, 2 |
302 |
|
AK050554.1 |
- |
1, 2 |
263 |
|
AK077695.1 |
- |
1, 2 |
265 |
|
AK082803.1 |
- |
1, 2 |
200 |
|
AK086448.1 |
- |
1, 2 |
338 |
|
AK086632.1 |
- |
1, 2 |
313 |
|
AK145399.1 |
- |
1, 2 |
299 |
|
AK166018.1 |
- |
1, 2 |
227 |
|
AK182882.1 |
- |
1, 2 |
57 |
|
AK215316.1 |
- |
1, 2 |
57 |
|
BC025803.1 |
- |
1, 2 |
312 |
5100552 |
View Full Product Details