Gene Transcripts

Gene Symbol
Hoxa6
Entrez Gene ID
15403
Gene Name
homeobox A6
Gene Aliases
Hox-1.2
Chromosome Location
Chr. 6: 52206365 - 52208624 on Build GRCm38
UniGene ID
Mm.222030
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010454.1 NP_034584.1 1 188
GenBank mRNA AB221546.1 - 1 188
BC119105.1 - 1 286 8733854
BC119107.1 - 1 286 8733856

View Full Product Details