Gene Transcripts
- Gene Symbol
- M6pr
- Entrez Gene ID
- 17113
- Gene Name
- mannose-6-phosphate receptor, cation dependent
- Gene Aliases
- CD-MPR, Mpr46
- Chromosome Location
- Chr. 6: 122309010 - 122317677 on Build GRCm38
- UniGene ID
- Mm.249225
- Species
- Mus musculus
- Gene Symbol
- M6pr-ps
- Entrez Gene ID
- 17114
- Gene Name
- mannose-6-phosphate receptor, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 3: 99500370 - 99502608 on Build GRCm38
- UniGene ID
- Mm.297872
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X64069.1 |
- |
1 |
1187 |
|
View Full Product Details