Gene Transcripts

Gene Symbol
Fanci
Entrez Gene ID
208836
Gene Name
Fanconi anemia, complementation group I
Gene Aliases
Chromosome Location
Chr. 7: 79392312 - 79450265 on Build GRCm38
UniGene ID
Mm.349493
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145946.2 NP_666058.2 23 2453
GenBank mRNA AK085572.1 - 21 2303
BC027836.1 - 21 2180

View Full Product Details