Gene Transcripts

Gene Symbol
Ttc28
Entrez Gene ID
209683
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
2310015L07Rik, 6030435N04, AI428795, AI851761, BC002262, TPRBK
Chromosome Location
Chr. 5: 110879784 - 111289780 on Build GRCm38
UniGene ID
Mm.295660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267622.1 NP_001254551.1 10 3557
GenBank mRNA AK122427.1 - 4 1120
BC046779.1 - 4 1221 5719497
BC060735.1 - 3 217

View Full Product Details