Gene Transcripts
- Gene Symbol
- Wnt7b
- Entrez Gene ID
- 22422
- Gene Name
- wingless-type MMTV integration site family, member 7B
- Gene Aliases
- Wnt-7b
- Chromosome Location
- Chr. 15: 85535437 - 85581821 on Build GRCm38
- UniGene ID
- Mm.306946
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC052018.1 |
- |
2 |
233 |
|
View Full Product Details