Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145832.3 | NP_665831.1 | 3 | 957 | |
NM_211358.2 | NP_997597.1 | 2 | 1445 | ||
GenBank mRNA | AJ440720.1 | - | 2 | 842 | |
AK030977.1 | - | 3 | 958 | ||
AK033295.1 | - | 2 | 534 | ||
AK040454.1 | - | 1 | 275 | ||
AK040986.1 | - | 2 | 1192 | ||
AK045972.1 | - | 3 | 934 | ||
AK050311.1 | - | 3 | 922 | ||
AK081068.1 | - | 2 | 943 | ||
AK088227.1 | - | 3 | 944 | ||
AK138595.1 | - | 2 | 1388 | ||
AK144762.1 | - | 2 | 1367 | ||
AK151154.1 | - | 2 | 1441 | ||
BC031814.1 | - | 2 | 1039 | 4196818 | |
BC116736.1 | - | 2 | 870 | 40126055 | |
BC119128.1 | - | 2 | 870 | 8733877 |